2-Amino-2-ethylbutanenitrile 98%
Supplier: Apollo Scientific
OR900555-1G OR900555-250MG
APOSOR900555-1GEA 364 EUR
APOSOR900555-1G APOSOR900555-250MG
2-Amino-2-ethylbutanenitrile 98%
2-Amino-2-ethylbutanenitrile
Formula: C₆H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD09730785
CAS Number: 22374-51-2