2,3-Dichloro-5-ethoxypyridine 98%
Supplier: Apollo Scientific
OR901903-1G OR901903-5G
APOSOR901903-1GEA 168 EUR
APOSOR901903-1G APOSOR901903-5G
2,3-Dichloro-5-ethoxypyridine 98%
2,3-Dichloro-5-ethoxypyridine
Formula: C₇H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD20441926
CAS Number: 1345471-27-3