1-(Pyridin-2-yl)-1H-pyrrole-2-carbaldehyde
Supplier: Apollo Scientific
OR902263-1G OR902263-250MG
APOSOR902263-1GEA 522 EUR
APOSOR902263-1G APOSOR902263-250MG
1-(Pyridin-2-yl)-1H-pyrrole-2-carbaldehyde
1-(Pyridin-2-yl)-1H-pyrrole-2-carbaldehyde
Formula: C₁₀H₈N₂O
MW: 172.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664911
CAS Number: 383136-44-5