Ethyl 5-cyclopropyl-1H-pyrazole-4-carboxylate 95%
Supplier: Apollo Scientific
OR902300-1G OR902300-5G
APOSOR902300-1GEA 74.1 EUR
APOSOR902300-1G APOSOR902300-5G
Ethyl 5-cyclopropyl-1H-pyrazole-4-carboxylate 95%
Ethyl 5-cyclopropyl-1H-pyrazole-4-carboxylate
Formula: C₉H₁₂N₂O₂
MW: 180.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753192
CAS Number: 1246471-38-4