(Cyclopropylmethyl)methylamine hydrochloride 98%
Supplier: Apollo Scientific
Synonyms: N-(Cyclopropylmethyl)-N-methylamine Hydrochloride
OR902516-1G OR902516-5G
APOSOR902516-1GEA 56.7 EUR
APOSOR902516-1G APOSOR902516-5G
(Cyclopropylmethyl)methylamine hydrochloride 98%
(Cyclopropylmethyl)methylamine hydrochloride
Formula: C₅H₁₁N·ClH
MW: 121.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759080
CAS Number: 77335-18-3