6-Methoxybenzofuran-2-carboxylic acid 98%
Supplier: Apollo Scientific
OR902576-1G
APOSOR902576-1GEA 72.5 EUR
APOSOR902576-1G
6-Methoxybenzofuran-2-carboxylic acid 98%
6-Methoxybenzofuran-2-carboxylic acid
Formula: C₁₀H₈O₄
MW: 192.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254721
CAS Number: 50551-61-6