2-Butylpiperidine hydrochloride 95%
Supplier: Apollo Scientific
OR903593-100MG
OR903593-1G
OR903593-250MG
APOSOR903593-100MGEA
190
EUR
APOSOR903593-100MG
APOSOR903593-1G
APOSOR903593-250MG
2-Butylpiperidine hydrochloride 95%
2-Butylpiperidine hydrochloride
Formula:
C₉H₁₉N·ClH MW: 177.715 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08277054 CAS Number: 72939-23-2 |