2-methyl-4H,5H,6H-cyclopenta[c]pyrazol-3-amine 98%
Supplier: Apollo Scientific
OR903604-1G OR903604-250MG OR903604-5G
APOSOR903604-1GEA 270 EUR
APOSOR903604-1G APOSOR903604-250MG APOSOR903604-5G
2-methyl-4H,5H,6H-cyclopenta[c]pyrazol-3-amine 98%
2-methyl-4H,5H,6H-cyclopenta[c]pyrazol-3-amine
Formula: C₇H₁₁N₃
MW: 137.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08444199
CAS Number: 877041-43-5