4-Ethoxymethylphenylboronic acid 97%
Supplier: Apollo Scientific
Synonyms:
4-(Ethoxymethyl)phenylboronic acid
OR903985-1G
OR903985-5G
APOSOR903985-1GEA
219
EUR
APOSOR903985-1G
APOSOR903985-5G
4-Ethoxymethylphenylboronic acid 97%
4-Ethoxymethylphenylboronic acid
Formula:
C₉H₁₃BO₃ MW: 180.009 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08701751 CAS Number: 279262-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...