5-Ethylindole-2-carboxylic acid 98%
Supplier: Apollo Scientific
OR904579-1G OR904579-5G
APOSOR904579-1GEA 190 EUR
APOSOR904579-1G APOSOR904579-5G
5-Ethylindole-2-carboxylic acid 98%
5-Ethylindole-2-carboxylic acid
Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065293
CAS Number: 37033-93-5