2,8-Diazaspiro[4.5]decan-1-one hydrochloride 97%
Supplier: Apollo Scientific
Warning
OR905065-1G OR905065-250MG OR905065-5G
APOSOR905065-1GEA 63.8 EUR
APOSOR905065-1G APOSOR905065-250MG APOSOR905065-5G
2,8-Diazaspiro[4.5]decan-1-one hydrochloride 97%
2,8-Diazaspiro[4.5]decan-1-one hydrochloride
Formula: C₈H₁₄N₂O·ClH
MW: 190.671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179151
CAS Number: 832710-65-3