2-(3,4-Dimethoxyphenyl)ethylisocyanide 95%
Supplier: Apollo Scientific
OR905902-1G OR905902-250MG
APOSOR905902-1GEA 683 EUR
APOSOR905902-1G APOSOR905902-250MG
2-(3,4-Dimethoxyphenyl)ethylisocyanide 95%
2-(3,4-Dimethoxyphenyl)ethylisocyanide
Formula: C₁₁H₁₃NO₂
MW: 191.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664626
CAS Number: 63609-01-8