N-tert-Butylpiperidine-4-carboxamide 98%
Supplier: Apollo Scientific
OR907366-1G OR907366-250MG
APOSOR907366-1GEA 574 EUR
APOSOR907366-1G APOSOR907366-250MG
N-tert-Butylpiperidine-4-carboxamide 98%
N-tert-Butylpiperidine-4-carboxamide
Formula: C₁₀H₂₀N₂O
MW: 184.279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739717
CAS Number: 86542-86-1