Isopropyl 3-cyanobenzoate 98%
Supplier: Apollo Scientific
OR907618-1G OR907618-5G
APOSOR907618-1GEA 219 EUR
APOSOR907618-1G APOSOR907618-5G
Isopropyl 3-cyanobenzoate 98%
Isopropyl 3-cyanobenzoate
Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981536
CAS Number: 110893-37-3