methyl 3-formyl-5-methoxybenzoate 98%
Supplier: Apollo Scientific
OR907907-1G OR907907-250MG
APOSOR907907-1GEA 444 EUR
APOSOR907907-1G APOSOR907907-250MG
methyl 3-formyl-5-methoxybenzoate 98%
methyl 3-formyl-5-methoxybenzoate
Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045770
CAS Number: 69026-10-4