Methyl 2-chloro-5-hydroxybenzoate 97%
Supplier: Apollo Scientific
OR907999-1G OR907999-250MG OR907999-5G
APOSOR907999-1GEA 149 EUR
APOSOR907999-1G APOSOR907999-250MG APOSOR907999-5G
Methyl 2-chloro-5-hydroxybenzoate 97%
Methyl 2-chloro-5-hydroxybenzoate
Formula: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698699
CAS Number: 247092-10-0