3-t-Butyl-1-(2-hydroxyethyl)pyrazole 97%
Supplier: Apollo Scientific
OR909281-1G OR909281-5G
APOSOR909281-1GEA 219 EUR
APOSOR909281-1G APOSOR909281-5G
3-t-Butyl-1-(2-hydroxyethyl)pyrazole 97%
3-t-Butyl-1-(2-hydroxyethyl)pyrazole
Formula: C₉H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD17214256
CAS Number: 1255574-70-9