1,5,6,7-Tetrahydro-2H-cyclopenta[b]pyridin-2-one 98%
Supplier: Apollo Scientific
OR909419-1G OR909419-5G
APOSOR909419-1GEA 219 EUR
APOSOR909419-1G APOSOR909419-5G
1,5,6,7-Tetrahydro-2H-cyclopenta[b]pyridin-2-one 98%
1,5,6,7-Tetrahydro-2H-cyclopenta[b]pyridin-2-one
Formula: C₈H₉NO
MW: 135.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09342899
CAS Number: 88499-85-8