1-(4-methylphenyl)ethanamine hydrochloride 98%
Supplier: Apollo Scientific
OR909505-100G OR909505-25G
APOSOR909505-100GEA 435 EUR
APOSOR909505-100G APOSOR909505-25G
1-(4-methylphenyl)ethanamine hydrochloride 98%
1-(4-methylphenyl)ethanamine hydrochloride
Formula: C₉H₁₃N·ClH
MW: 171.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22383689
CAS Number: 854181-94-5