2-Methoxy-4-morpholinopyrimidine 98%
Supplier: Apollo Scientific
OR909557-1G OR909557-5G
APOSOR909557-1GEA 219 EUR
APOSOR909557-1G APOSOR909557-5G
2-Methoxy-4-morpholinopyrimidine 98%
2-Methoxy-4-morpholinopyrimidine
Formula: C₉H₁₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD22421507
CAS Number: 1400644-32-7