1-Propylpyrazol-4-amine hydrochloride 98%
Supplier: Apollo Scientific
OR909656-1G OR909656-5G
APOSOR909656-1GEA 168 EUR
APOSOR909656-1G APOSOR909656-5G
1-Propylpyrazol-4-amine hydrochloride 98%
1-Propylpyrazol-4-amine hydrochloride
Formula: C₆H₁₁N₃·ClH
MW: 161.633 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22666476
CAS Number: 1390654-83-7