2-Methyl-5-(4-methylphenyl)pyrazol-3-amine 98%
Supplier: Apollo Scientific
Synonyms: 1-Methyl-3-(p-tolyl)-1H-pyrazol-5-amine
OR909717-10G OR909717-1G OR909717-5G
APOSOR909717-10GEA 754 EUR
APOSOR909717-10G APOSOR909717-1G APOSOR909717-5G
2-Methyl-5-(4-methylphenyl)pyrazol-3-amine 98%
2-Methyl-5-(4-methylphenyl)pyrazol-3-amine
Formula: C₁₁H₁₃N₃
MW: 187.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06637391
CAS Number: 126417-86-5