5-Cyclopropyl-2-methyl-1H-pyrazol-3-one 96%
Supplier: Apollo Scientific
OR909744-1G OR909744-5G
APOSOR909744-1GEA 140 EUR
APOSOR909744-1G APOSOR909744-5G
5-Cyclopropyl-2-methyl-1H-pyrazol-3-one 96%
5-Cyclopropyl-2-methyl-1H-pyrazol-3-one
Formula: C₇H₁₀N₂O
MW: 138.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21667125
CAS Number: 199125-21-8