5-Chloro-4-methoxy-2-methylaniline 97%
Supplier: Apollo Scientific
OR909849-1G OR909849-5G
APOSOR909849-1GEA 190 EUR
APOSOR909849-1G APOSOR909849-5G
5-Chloro-4-methoxy-2-methylaniline 97%
5-Chloro-4-methoxy-2-methylaniline
Formula: C₈H₁₀ClNO
MW: 171.624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783103
CAS Number: 62492-46-0