6-Bromoimidazo[1,5-a]pyridine 95%
Supplier: Apollo Scientific
Warning
OR910029-1G OR910029-250MG
APOSOR910029-1GEA 294 EUR
APOSOR910029-1G APOSOR910029-250MG
6-Bromoimidazo[1,5-a]pyridine 95%
6-Bromoimidazo[1,5-a]pyridine
Formula: C₇H₅BrN₂
Melting Pt: 240…246 °C
Storage Temperature: Ambient
MDL Number: MFCD17677174
CAS Number: 1239880-00-2