Methyl 2-(1H-pyrazol-1-yl)acetate 98%
Supplier: Apollo Scientific
OR910074-1G OR910074-250MG OR910074-5G
APOSOR910074-1GEA 78.3 EUR
APOSOR910074-1G APOSOR910074-250MG APOSOR910074-5G
Methyl 2-(1H-pyrazol-1-yl)acetate 98%
Methyl 2-(1H-pyrazol-1-yl)acetate
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02029025
CAS Number: 142890-12-8