1-{[2-(trimethylsilyl)ethoxy]methyl}pyrazole 95%
Supplier: Apollo Scientific
Synonyms: 1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazole
OR910093-1G OR910093-250MG OR910093-5G
APOSOR910093-1GEA 109 EUR
APOSOR910093-1G APOSOR910093-250MG APOSOR910093-5G
1-{[2-(trimethylsilyl)ethoxy]methyl}pyrazole 95%
1-{[2-(trimethylsilyl)ethoxy]methyl}pyrazole
Formula: C₉H₁₈N₂OSi
Storage Temperature: Ambient
MDL Number: MFCD12405511
CAS Number: 133560-57-3