2-N-Isopropylamino-5-nitropyridine 98%
Supplier: Apollo Scientific
OR911055-1G OR911055-5G
APOSOR911055-1GEA 161 EUR
APOSOR911055-1G APOSOR911055-5G
2-N-Isopropylamino-5-nitropyridine 98%
2-N-Isopropylamino-5-nitropyridine
Formula: C₈H₁₁N₃O₂
MW: 181.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11116818
CAS Number: 26820-53-1