N-Isopropylthiophene-3-carboxamide 98%

Supplier: Apollo Scientific

OR911186-1G OR911186-5G
APOSOR911186-1GEA 270 EUR
APOSOR911186-1G APOSOR911186-5G
N-Isopropylthiophene-3-carboxamide 98%
N-Isopropylthiophene-3-carboxamide
Formula: C₈H₁₁NOS
MW: 169.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10672923
CAS Number: 531549-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR