5-Nitro-2-indanone 98%
Supplier: Apollo Scientific
Synonyms: 5-Nitro-1H-inden-2(3H)-one
OR912539-1G OR912539-5G
APOSOR912539-1GEA 92.8 EUR
APOSOR912539-1G APOSOR912539-5G
5-Nitro-2-indanone 98%
5-Nitro-2-indanone
Formula: C₉H₇NO₃
MW: 177.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00598938
CAS Number: 116530-60-0