2,3-Dihydroindol-5-ol 95%
Supplier: Apollo Scientific
Synonyms: Indolin-5-ol
OR912750-1G OR912750-250MG
APOSOR912750-1GEA 155 EUR
APOSOR912750-1G APOSOR912750-250MG
2,3-Dihydroindol-5-ol 95%
2,3-Dihydroindol-5-ol
Formula: C₈H₉NO
MW: 135.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02181134
CAS Number: 172078-33-0