7-Methoxy-3-indolecarboxaldehyde 98%
Supplier: Apollo Scientific
OR912786-1G OR912786-5G
APOSOR912786-1GEA 139 EUR
APOSOR912786-1G APOSOR912786-5G
7-Methoxy-3-indolecarboxaldehyde 98%
7-Methoxy-3-indolecarboxaldehyde
Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02667609
CAS Number: 109021-59-2