(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanecarboxylic acid 95%
Supplier: Apollo Scientific
OR913229-1G OR913229-25G OR913229-5G
APOSOR913229-1GEA 64.4 EUR
APOSOR913229-1G APOSOR913229-25G APOSOR913229-5G
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanecarboxylic acid 95%
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanecarboxylic acid
Formula: C₁₁H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00798812
CAS Number: 16052-40-7