(R)-(1-Benzyl-pyrrolidin-3-yl)-methanol 95%
Supplier: Apollo Scientific
OR913599-1G
OR913599-250MG
APOSOR913599-1GEA
806
EUR
APOSOR913599-1G
APOSOR913599-250MG
(R)-(1-Benzyl-pyrrolidin-3-yl)-methanol 95%
(R)-(1-Benzyl-pyrrolidin-3-yl)-methanol
Formula:
C₁₂H₁₇NO MW: 191.269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09608007 CAS Number: 303111-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...