(S)-Tetrahydro-2H-pyran-3-amine hydrochloride 95%
Supplier: Apollo Scientific
OR914019-1G OR914019-250MG
APOSOR914019-1GEA 206 EUR
APOSOR914019-1G APOSOR914019-250MG
(S)-Tetrahydro-2H-pyran-3-amine hydrochloride 95%
(S)-Tetrahydro-2H-pyran-3-amine hydrochloride
Formula: C₅H₁₁NO·ClH
MW: 137.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09056862
CAS Number: 1071829-81-6