[1,2,4]Triazolo[1,5-a]pyridin-2-amine 95%
Supplier: Apollo Scientific
OR914046-100MG OR914046-1G OR914046-250MG
APOSOR914046-100MGEA 29 EUR
APOSOR914046-100MG APOSOR914046-1G APOSOR914046-250MG
[1,2,4]Triazolo[1,5-a]pyridin-2-amine 95%
[1,2,4]Triazolo[1,5-a]pyridin-2-amine
Formula: C₆H₆N₄
MW: 134.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846603
CAS Number: 874-46-4