1-(1H-Pyrazol-1-yl)ethanone 95%
Supplier: Apollo Scientific
OR914063-1G OR914063-250MG
APOSOR914063-1GEA 487 EUR
APOSOR914063-1G APOSOR914063-250MG
1-(1H-Pyrazol-1-yl)ethanone 95%
1-(1H-Pyrazol-1-yl)ethanone
Formula: C₅H₆N₂O
MW: 110.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01336293
CAS Number: 10199-64-1