1-(1H-Pyrrolo[3,2-b]pyridin-3-yl)ethanone 95%
Supplier: Apollo Scientific
OR914064-1G OR914064-250MG
APOSOR914064-1GEA 886 EUR
APOSOR914064-1G APOSOR914064-250MG
1-(1H-Pyrrolo[3,2-b]pyridin-3-yl)ethanone 95%
1-(1H-Pyrrolo[3,2-b]pyridin-3-yl)ethanone
Formula: C₉H₈N₂O
MW: 160.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834630
CAS Number: 460053-62-7