1-(3-Chloropyrazin-2-yl)ethanone 95%
Supplier: Apollo Scientific
Warning
OR914097-1G OR914097-250MG
APOSOR914097-1GEA 125 EUR
APOSOR914097-1G APOSOR914097-250MG
1-(3-Chloropyrazin-2-yl)ethanone 95%
1-(3-Chloropyrazin-2-yl)ethanone
Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697755
CAS Number: 121246-90-0