2-Bromo-5-cyano-6-picoline 97%
Supplier: Apollo Scientific
Synonyms: 6-Bromo-2-methylnicotinonitrile
OR914314-1G OR914314-5G
APOSOR914314-1GEA 133 EUR
APOSOR914314-1G APOSOR914314-5G
2-Bromo-5-cyano-6-picoline 97%
2-Bromo-5-cyano-6-picoline
Formula: C₇H₅BrN₂
MW: 197.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839266
CAS Number: 1003711-39-4