Indoline-5-carboxylic acid hydrochloride 95%
Supplier: Apollo Scientific
OR916518-1G OR916518-250MG
APOSOR916518-1GEA 806 EUR
APOSOR916518-1G APOSOR916518-250MG
Indoline-5-carboxylic acid hydrochloride 95%
Indoline-5-carboxylic acid hydrochloride
Formula: C₉H₉NO₂·ClH
MW: 199.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971545
CAS Number: 1171876-52-0