2-Chloro-6-methoxybenzamide 95%
Supplier: Apollo Scientific
OR916916-1G OR916916-250MG
APOSOR916916-1GEA 435 EUR
APOSOR916916-1G APOSOR916916-250MG
2-Chloro-6-methoxybenzamide 95%
2-Chloro-6-methoxybenzamide
Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846167
CAS Number: 107485-43-8