1-(1-Hydroxy-cyclopentyl)-ethanone 95%
Supplier: Apollo Scientific
OR916980-1G OR916980-250MG
APOSOR916980-1GEA 632 EUR
APOSOR916980-1G APOSOR916980-250MG
1-(1-Hydroxy-cyclopentyl)-ethanone 95%
1-(1-Hydroxy-cyclopentyl)-ethanone
Formula: C₇H₁₂O₂
MW: 128.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658286
CAS Number: 17160-89-3