Thieno[2,3-c]pyridine-2-carbaldehyde 95%
Supplier: Apollo Scientific
Warning
OR917276-1G OR917276-250MG
APOSOR917276-1GEA 649 EUR
APOSOR917276-1G APOSOR917276-250MG
Thieno[2,3-c]pyridine-2-carbaldehyde 95%
Thieno[2,3-c]pyridine-2-carbaldehyde
Formula: C₈H₅NOS
MW: 163.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052828
CAS Number: 203922-18-3