2,7-Diazaspiro[4.5]decan-3-one 95+%
Supplier: Apollo Scientific
Warning
OR917833-100MG OR917833-250MG
APOSOR917833-100MGEA 108 EUR
APOSOR917833-100MG APOSOR917833-250MG
2,7-Diazaspiro[4.5]decan-3-one 95+%
2,7-Diazaspiro[4.5]decan-3-one
Formula: C₈H₁₄N₂O
MW: 154.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12406549
CAS Number: 1158750-89-0