Fmoc-L-Arg(Me)2(Pbf)-OH
Supplier: Apollo Scientific
Warning
Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(((dimethylamino)((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran)-5-sulphonamido)methylene)amino)pentanoic acid
OR917976-1G OR917976-250MG
APOSOR917976-1GEA 864 EUR
APOSOR917976-1G APOSOR917976-250MG
Fmoc-L-Arg(Me)2(Pbf)-OH
Fmoc-L-Arg(Me)2(Pbf)-OH
Formula: C₃₆H₄₄N₄O₇S
MW: 676.822 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15141977
CAS Number: 1185841-84-2