8-Methyl-3,4-dihydro-2H-1-benzopyran-4-one 95%

Supplier: Apollo Scientific

Synonyms: 8-Methyl-4-chromanone

OR918210-1G OR918210-250MG
APOSOR918210-1GEA 294 EUR
APOSOR918210-1G APOSOR918210-250MG
8-Methyl-3,4-dihydro-2H-1-benzopyran-4-one 95%
8-Methyl-3,4-dihydro-2H-1-benzopyran-4-one
Formula: C₁₀H₁₀O₂
MW: 162.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054820
CAS Number: 49660-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR