5-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 98%
Supplier: Apollo Scientific
OR918489-1G OR918489-250MG
APOSOR918489-1GEA 112 EUR
APOSOR918489-1G APOSOR918489-250MG
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 98%
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formula: C₇H₃ClN₄
MW: 178.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18073765
CAS Number: 1224288-92-9