4-(5-Formylpyridin-2-yl)piperazine, N1-BOC protected 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate, 2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]-5-formylpyridine
OR9195-1G OR9195-100MG OR9195-250MG
APOSOR9195-1GEA 331 EUR
APOSOR9195-1G APOSOR9195-100MG APOSOR9195-250MG
4-(5-Formylpyridin-2-yl)piperazine, N1-BOC protected 97%
4-(5-Formylpyridin-2-yl)piperazine, N1-BOC protected
Formula: C₁₅H₂₁N₃O₃
MW: 291.346 g/mol
Melting Pt: 122…128 °C
Storage Temperature: Ambient
MDL Number: MFCD08271923
CAS Number: 479226-10-3