Methyl 2-(1-methyl-2-pyrrolidylidene)acetate 95%
Supplier: Apollo Scientific
OR919522-1G OR919522-250MG
APOSOR919522-1GEA 435 EUR
APOSOR919522-1G APOSOR919522-250MG
Methyl 2-(1-methyl-2-pyrrolidylidene)acetate 95%
Methyl 2-(1-methyl-2-pyrrolidylidene)acetate
Formula: C₈H₁₃NO₂
MW: 155.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381105
CAS Number: 78167-68-7